CAS RN 4333-20-4
CAS RN 4333-20-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Phenylthiohydantoin-valine 98.0+%, TCI Americaâ„¢
CAS: 4333-20-4 Molecular Formula: C12H14N2OS Molecular Weight (g/mol): 234.32 MDL Number: MFCD00038438 InChI Key: HEIKHJHSVLQGGN-UHFFFAOYNA-N Synonym: 5-Isopropyl-3-phenyl-2-thiohydantoin, PTH-valine PubChem CID: 2831422 IUPAC Name: 3-phenyl-5-(propan-2-yl)-2-sulfanylideneimidazolidin-4-one SMILES: CC(C)C1NC(=S)N(C1=O)C1=CC=CC=C1