accessibility menu, dialog, popup

UserName

CAS RN 433-94-3

IUPAC Name: 2-chloro-6-(trifluoromethyl)aniline
Synonyms: 2-chloro-6-(trifluoromethyl)aniline
Molecular Weight (g/mol): 195.57
Molecular Formula: C7H5ClF3N
InChi Key: OTRRSPQJZRCMDA-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=CC=C1C(F)(F)F
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)