missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-(trifluoromethyl)aniline, 97%

Catalog No. AAB2271414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2271414 25 g
AAB2271406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2271414 Supplier Thermo Scientific Chemicals Supplier No. B2271414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 433-94-3 | C7H5ClF3N | 195.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 433-94-3
Molecular Formula C7H5ClF3N
Molecular Weight (g/mol) 195.57
MDL Number MFCD00272565
InChI Key OTRRSPQJZRCMDA-UHFFFAOYSA-N
Synonym 2-chloro-6-trifluoromethyl aniline, 2-amino-3-chlorobenzotrifluoride, benzenamine, 2-chloro-6-trifluoromethyl, 2-chloranyl-6-trifluoromethyl aniline, pubchem2764, 2-chloro-6-trifluoromethyl benzenamine, 2-chloro-6-trifluoromethyl phenylamine, 6-chloro-2-trifluoromethyl phenylamine, 2-chloro-6-trifluoromethyl-benzenamine, 2-chloro-6-trifluoromethyl aniline 2-amino-3-chlorobenzotrifluoride
PubChem CID 2756384
IUPAC Name 2-chloro-6-(trifluoromethyl)aniline
SMILES NC1=C(Cl)C=CC=C1C(F)(F)F

Specifications

Density 1.42
Boiling Point 39°C to 40°C (0.1 mmHg)
Quantity 25 g
UN Number UN2810
Formula Weight 195.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-(trifluoromethyl)aniline
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 207-091-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only