missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-chlorobenzotrifluoride 97.0+%, TCI America™

Catalog No. A15105G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15105G 5 g
A151025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15105G Supplier TCI America Supplier No. A15105G

Chemical Identifiers

CAS 433-94-3
Molecular Formula C7H5ClF3N
Molecular Weight (g/mol) 195.57
MDL Number MFCD00272565
InChI Key OTRRSPQJZRCMDA-UHFFFAOYSA-N
Synonym 2-chloro-6-trifluoromethyl aniline, 2-amino-3-chlorobenzotrifluoride, benzenamine, 2-chloro-6-trifluoromethyl, 2-chloranyl-6-trifluoromethyl aniline, pubchem2764, 2-chloro-6-trifluoromethyl benzenamine, 2-chloro-6-trifluoromethyl phenylamine, 6-chloro-2-trifluoromethyl phenylamine, 2-chloro-6-trifluoromethyl-benzenamine, 2-chloro-6-trifluoromethyl aniline 2-amino-3-chlorobenzotrifluoride
PubChem CID 2756384
IUPAC Name 2-chloro-6-(trifluoromethyl)aniline
SMILES NC1=C(Cl)C=CC=C1C(F)(F)F

Specifications

Color Yellow
Boiling Point 40°C
Quantity 5 g
Formula Weight 195.57
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Amino-3-chlorobenzotrifluoride
EINECSNumber (3)-0266
TSCA No