accessibility menu, dialog, popup

UserName

CAS RN 433-53-4

IUPAC Name: methyl 2,2-difluoroacetate
Synonyms: methyl 2,2-difluoroacetate
Molecular Weight (g/mol): 110.06
Molecular Formula: C3H4F2O2
InChi Key: CSSYKHYGURSRAZ-UHFFFAOYSA-N
SMILES: COC(=O)C(F)F
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)