accessibility menu, dialog, popup

UserName

H3COHOH3C

CAS RN 4317-61-7

IUPAC Name: 1-(2-methylphenoxy)propan-2-ol
Synonyms: 2-propanol, 1-(o-tolyloxy)-
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: ISIQEWJJVXFCDP-UHFFFAOYNA-N
SMILES: CC(O)COC1=CC=CC=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results