missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Methylphenoxy)-2-propanol 90.0+%, TCI America™

Catalog No. M10635G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M10635G Supplier TCI America Supplier No. M10635G
Only null left

Chemical Identifiers

CAS 4317-61-7
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00191542
InChI Key ISIQEWJJVXFCDP-UHFFFAOYSA-N
Synonym 1-(o-Tolyloxy)-2-propanol
PubChem CID 107264
IUPAC Name 1-(2-methylphenoxy)propan-2-ol
SMILES CC1=CC=CC=C1OCC(C)O

Specifications

Color Yellow
Boiling Point 130°C
Quantity 5 g
Formula Weight 166.22
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(2-Methylphenoxy)-2-propanol
RTECSNumber UB9650000
TSCA No