accessibility menu, dialog, popup

UserName

HClNH

CAS RN 43064-12-6

IUPAC Name: hydrogen 4-phenyl-1,2,3,6-tetrahydropyridine chloride
Synonyms: hydrogen 4-phenyl-1,2,3,6-tetrahydropyridine chloride
Molecular Weight (g/mol): 195.69
Molecular Formula: C11H14ClN
InChi Key: POGWXTJNUCZEPR-UHFFFAOYSA-N
SMILES: [H+].[Cl-].C1CC(=CCN1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)