CAS RN 42797-18-2
IUPAC Name:
2-{[1,1'-biphenyl]-4-carbonyl}benzoic acid
Synonyms:
2-{[1,1'-biphenyl]-4-carbonyl}benzoic acid
Molecular Weight (g/mol):
302.33
Molecular Formula:
C20H14O3
InChi Key:
NBYNVBRXIUUSIG-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=CC=C1C(=O)C1=CC=C(C=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
2-(4-Biphenylylcarbonyl)benzoic Acid 97.0+%, TCI America™
CAS: 42797-18-2 Molecular Formula: C20H14O3 Molecular Weight (g/mol): 302.33 MDL Number: MFCD00392368 InChI Key: NBYNVBRXIUUSIG-UHFFFAOYSA-N PubChem CID: 39302 IUPAC Name: 2-{[1,1'-biphenyl]-4-carbonyl}benzoic acid SMILES: OC(=O)C1=CC=CC=C1C(=O)C1=CC=C(C=C1)C1=CC=CC=C1