missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Biphenylylcarbonyl)benzoic Acid 97.0+%, TCI America™

Catalog No. B45325G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45325G 5 g
B45321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45325G Supplier TCI America Supplier No. B45325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 42797-18-2
Molecular Formula C20H14O3
Molecular Weight (g/mol) 302.33
MDL Number MFCD00392368
InChI Key NBYNVBRXIUUSIG-UHFFFAOYSA-N
PubChem CID 39302
IUPAC Name 2-{[1,1'-biphenyl]-4-carbonyl}benzoic acid
SMILES OC(=O)C1=CC=CC=C1C(=O)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 229°C
Color White
Quantity 5 g
Formula Weight 302.33
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Biphenylylcarbonyl)benzoic Acid
RTECSNumber DG4375000
TSCA Yes