accessibility menu, dialog, popup

UserName

CAS RN 42712-64-1

IUPAC Name: 2-amino-1,4-dihydroquinolin-4-one
Synonyms: 2-amino-1H-quinolin-4-one
Molecular Weight (g/mol): 160.18
Molecular Formula: C9H8N2O
InChi Key: LWGUCIXHBVVATR-UHFFFAOYSA-N
SMILES: NC1=CC(=O)C2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results