missing translation for 'onlineSavingsMsg'
Learn More

Methyl Isobutyrylacetate 97.0+%, TCI America™

Catalog No. I059225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I059225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I059225G Supplier TCI America Supplier No. I059225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 42558-54-3
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00040499
InChI Key HNNFDXWDCFCVDM-UHFFFAOYSA-N
Synonym methyl isobutyrylacetate, methyl 4-methyl-3-oxovalerate, 4-methyl-3-oxovaleric acid methyl ester, iso-butyryl methyl acetate, methyl 4-methyl-3-oxo-pentanoate, methylisobutyrylacetate, acmc-209jq9, ksc237g2d, methyl 4-methyl-3oxopentanoate, methyl4-methyl-3-oxopentanoate
PubChem CID 2759969
IUPAC Name methyl 4-methyl-3-oxopentanoate
SMILES CC(C)C(=O)CC(=O)OC

Specifications

Color Yellow
Quantity 25 g
Formula Weight 144.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Isobutyrylacetate