accessibility menu, dialog, popup

UserName

CAS RN 42389-50-4

IUPAC Name: N1,N1-dimethylcyclohexane-1,4-diamine
Synonyms: N1,N1-dimethylcyclohexane-1,4-diamine
Molecular Weight (g/mol): 142.25
Molecular Formula: C8H18N2
InChi Key: GNGZZWTXFNFCMU-UHFFFAOYSA-N
SMILES: CN(C)C1CCC(N)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results