accessibility menu, dialog, popup

UserName

H3COOHO

CAS RN 42058-59-3

IUPAC Name: methyl 2-(3-hydroxyphenyl)acetate
Synonyms: methyl 2-(3-hydroxyphenyl)acetate
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: AMDDOQIUPAINLH-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC=CC(O)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results