missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Hydroxyphenylacetate 98.0+%, TCI America™

Catalog No. M29255G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29255G 5 g
M29251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29255G Supplier TCI America Supplier No. M29255G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 42058-59-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00017660
InChI Key AMDDOQIUPAINLH-UHFFFAOYSA-N
Synonym 3-Hydroxyphenylacetic Acid Methyl Ester
PubChem CID 39129
IUPAC Name methyl 2-(3-hydroxyphenyl)acetate
SMILES COC(=O)CC1=CC(=CC=C1)O

Specifications

Color Yellow
Boiling Point 146°C
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Hydroxyphenylacetate