accessibility menu, dialog, popup

UserName

CAS RN 42048-11-3

CH3ICl

CAS RN 42048-11-3

IUPAC Name: 1-chloro-3-iodo-2-methylbenzene
Synonyms: 1-chloro-3-iodo-2-methylbenzene
Molecular Weight (g/mol): 252.48
Molecular Formula: C7H6ClI
InChi Key: OEHHXVIJMCMYGM-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C=CC=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results