missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-iodotoluene 98.0+%, TCI America™

Catalog No. C16031G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C16031G Supplier TCI America Supplier No. C16031G
Only null left

Chemical Identifiers

CAS 42048-11-3
Molecular Formula C7H6ClI
Molecular Weight (g/mol) 252.479
MDL Number MFCD00019013
InChI Key OEHHXVIJMCMYGM-UHFFFAOYSA-N
Synonym 2-chloro-6-iodotoluene, 6-chloro-2-iodotoluene, 1-chloro-3-iodo-2-methyl-benzene, pubchem3654, 2-iodo-6-chlorotoluene, acmc-1aru0, ksc491k9p, 3-chloro-1-iodo-2-methylbenzene, buttpark 121\11-22, 3-chloro-2-methyliodobenzene
PubChem CID 2757617
IUPAC Name 1-chloro-3-iodo-2-methylbenzene
SMILES CC1=C(C=CC=C1I)Cl

Specifications

Boiling Point 133°C
Quantity 1 g
Formula Weight 252.48
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-6-iodotoluene
TSCA No
Recommended Storage Refrigerator