accessibility menu, dialog, popup

UserName

CAS RN 41859-67-0

IUPAC Name: 2-(4-{2-[(4-chlorophenyl)formamido]ethyl}phenoxy)-2-methylpropanoic acid
Synonyms: bezafibrate bezacur merckle brand of bezafibrate bezafibrate (substance) bezafibrato [inn-spanish] bezafibrate synthelabo brand reducterol befibrat betapharm brand of bezafibrate hennig brand of bezafibrate
Molecular Weight (g/mol): 361.82
Molecular Formula: C19H20ClNO4
InChi Key: IIBYAHWJQTYFKB-UHFFFAOYSA-N
SMILES: CC(C)(OC1=CC=C(CCNC(=O)C2=CC=C(Cl)C=C2)C=C1)C(O)=O
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)