missing translation for 'onlineSavingsMsg'
Learn More

Bezafibrate, 98+%

Catalog No. AAJ61412ME
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61412ME 500 mg
AAJ6141203 1 g
AAJ6141206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ61412ME Supplier Thermo Scientific Chemicals Supplier No. J61412ME

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A peroxisome proliferator-activated receptor agonist and hypolipidemic agent | CAS: 41859-67-0 | C19H20ClNO4 | 361.82 g/mol

A peroxisome proliferator-activated receptor agonist and hypolipidemic agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 41859-67-0
Molecular Formula C19H20ClNO4
Molecular Weight (g/mol) 361.82
MDL Number MFCD00078970
InChI Key IIBYAHWJQTYFKB-UHFFFAOYSA-N
Synonym bezafibrate, bezalip, cedur, bezafibrat, befizal, bezafibrato, bezafibratum, sklerofibrat, azufibrat, difaterol
PubChem CID 39042
ChEBI CHEBI:47612
IUPAC Name 2-(4-{2-[(4-chlorophenyl)formamido]ethyl}phenoxy)-2-methylpropanoic acid
SMILES CC(C)(OC1=CC=C(CCNC(=O)C2=CC=C(Cl)C=C2)C=C1)C(O)=O

Specifications

Melting Point 183°C to 186°C
Color White
Quantity 500 mg
Merck Index 14,1195
Formula Weight 361.82
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Bezafibrate
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 255-567-9
RTECSNumber UE8755000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only