missing translation for 'onlineSavingsMsg'
Learn More

Bezafibrate 98.0+%, TCI America™

Catalog No. B33465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33465G 5 g
B334625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33465G Supplier TCI America Supplier No. B33465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41859-67-0
Molecular Formula C19H20ClNO4
Molecular Weight (g/mol) 361.82
MDL Number MFCD00078970
InChI Key IIBYAHWJQTYFKB-UHFFFAOYSA-N
Synonym bezafibrate, bezalip, cedur, bezafibrat, befizal, bezafibrato, bezafibratum, sklerofibrat, azufibrat, difaterol
PubChem CID 39042
ChEBI CHEBI:47612
IUPAC Name 2-(4-{2-[(4-chlorophenyl)formamido]ethyl}phenoxy)-2-methylpropanoic acid
SMILES CC(C)(OC1=CC=C(CCNC(=O)C2=CC=C(Cl)C=C2)C=C1)C(O)=O

Specifications

Melting Point 184°C
Color White-Yellow
Quantity 5 g
Formula Weight 361.82
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bezafibrate
RTECSNumber UE8755000
TSCA No