accessibility menu, dialog, popup

UserName

CH3OOBrOHBr

CAS RN 41727-47-3

IUPAC Name: methyl 3,5-dibromo-4-hydroxybenzoate
Synonyms: methyl 3,5-dibromo-4-hydroxybenzoate
Molecular Weight (g/mol): 309.94
Molecular Formula: C8H6Br2O3
InChi Key: NVGJGYZKXBLIKY-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Br)=C(O)C(Br)=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)