missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-dibromo-4-hydroxybenzoate, 98%

Catalog No. AAA1079930
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1079930 250 g
AAA1079909 10 g
AAA1079918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1079930 Supplier Thermo Scientific Chemicals Supplier No. A1079930
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 41727-47-3 | C8H6Br2O3 | 309.941 g/mol

Methyl 3,5-dibromo-4-hydroxybenzoate is used to produce 4-hydroxy-3,5-di-pyridin-2-yl-benzoic acid methyl ester at temperature of 90-100°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3,5-dibromo-4-hydroxybenzoate is used to produce 4-hydroxy-3,5-di-pyridin-2-yl-benzoic acid methyl ester at temperature of 90-100°C.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 41727-47-3
Molecular Formula C8H6Br2O3
Molecular Weight (g/mol) 309.941
MDL Number MFCD00016416
InChI Key NVGJGYZKXBLIKY-UHFFFAOYSA-N
Synonym 3,5-dibromo-4-hydroxybenzoic acid methyl ester, 3,5-dibromo-4-hydroxy-benzoic acid methyl ester, methyl 3,5-bis bromanyl-4-oxidanyl-benzoate, acmc-20akhs, pubchem3978, methyl dibromosalicylate, rarechem al bf 0231, timtec-bb sbb012530, labotest-bb lt00455670, labotest-bb lt03332530
PubChem CID 726975
IUPAC Name methyl 3,5-dibromo-4-hydroxybenzoate
SMILES COC(=O)C1=CC(=C(C(=C1)Br)O)Br

Specifications

Melting Point 123°C to 125°C
Quantity 250 g
Beilstein 2373854
Solubility Information Slightly soluble in water.
Formula Weight 309.94
Percent Purity 98%
Chemical Name or Material Methyl 3,5-dibromo-4-hydroxybenzoate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 255-521-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only