accessibility menu, dialog, popup

UserName

CH3O(Z)

CAS RN 4170-30-3

IUPAC Name: (2Z)-but-2-enal
Synonyms: crotonaldehyde
Molecular Weight (g/mol): 70.09
Molecular Formula: C4H6O
InChi Key: MLUCVPSAIODCQM-IHWYPQMZSA-N
SMILES: C\C=C/C=O
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)