accessibility menu, dialog, popup

UserName

H3CSClClCl

CAS RN 4163-78-4

IUPAC Name: 1,2,4-trichloro-5-(methylsulfanyl)benzene
Synonyms: 1,2,4-trichloro-5-(methylsulfanyl)benzene
Molecular Weight (g/mol): 227.53
Molecular Formula: C7H5Cl3S
InChi Key: RWDHTEMSFGMSDP-UHFFFAOYSA-N
SMILES: CSC1=CC(Cl)=C(Cl)C=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results