accessibility menu, dialog, popup

UserName

CAS RN 41302-05-0

ClOO

CAS RN 41302-05-0

IUPAC Name: 2-(4-chlorobutoxy)oxane
Synonyms: 2-(4-chlorobutoxy)oxane
Molecular Weight (g/mol): 192.68
Molecular Formula: C9H17ClO2
InChi Key: SZZINZURZKQZLG-UHFFFAOYNA-N
SMILES: ClCCCCOC1CCCCO1
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results