accessibility menu, dialog, popup

UserName

CAS RN 4124-31-6

IUPAC Name: 2,2,2-trichloroacetyl 2,2,2-trichloroacetate
Synonyms: trichloroacetic anhydride
Molecular Weight (g/mol): 308.74
Molecular Formula: C4Cl6O3
InChi Key: MEFKFJOEVLUFAY-UHFFFAOYSA-N
SMILES: ClC(Cl)(Cl)C(=O)OC(=O)C(Cl)(Cl)Cl
Boiling Point 
  • (1)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)