missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic anhydride, 95%

Catalog No. AC446091000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC446091000 100 g
AC446090250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446091000 Supplier Thermo Scientific Chemicals Supplier No. 446091000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4124-31-6 | C4Cl6O3 | 308.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4124-31-6
Molecular Formula C4Cl6O3
Molecular Weight (g/mol) 308.76
InChI Key MEFKFJOEVLUFAY-UHFFFAOYSA-N
Synonym trichloroacetic anhydride, acetic acid, trichloro-, anhydride, acetic acid, 2,2,2-trichloro-, 1,1'-anhydride, acmc-1aliy, perchloroacetic anhydride, bis trichloroacetic anhydride, trichloroacetic acid anhydride, 2,2,2-trichloroacetic anhydride, trichloroacetyl 2,2,2-trichloroacetate
PubChem CID 20079
IUPAC Name (2,2,2-trichloroacetyl) 2,2,2-trichloroacetate
SMILES C(=O)(C(Cl)(Cl)Cl)OC(=O)C(Cl)(Cl)Cl

Specifications

Density 1.6900g/mL
Boiling Point 139°C to 141°C (60.0 mmHg)
Assay Percent Range 95%
Linear Formula [CCl3C(=O)]2O
Quantity 100 g
Specific Gravity 1.69
Solubility Information Solubility in water: reacts
Formula Weight 308.76
Percent Purity 95%
Chemical Name or Material Trichloroacetic anhydride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 223-925-3

RUO – Research Use Only