missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic Anhydride 97.0+%, TCI America™

Catalog No. T079225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T079225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T079225G Supplier TCI America Supplier No. T079225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4124-31-6
Molecular Formula C4Cl6O3
Molecular Weight (g/mol) 308.741
MDL Number MFCD00000793
InChI Key MEFKFJOEVLUFAY-UHFFFAOYSA-N
Synonym trichloroacetic anhydride, acetic acid, trichloro-, anhydride, acetic acid, 2,2,2-trichloro-, 1,1'-anhydride, acmc-1aliy, perchloroacetic anhydride, bis trichloroacetic anhydride, trichloroacetic acid anhydride, 2,2,2-trichloroacetic anhydride, trichloroacetyl 2,2,2-trichloroacetate
PubChem CID 20079
IUPAC Name (2,2,2-trichloroacetyl) 2,2,2-trichloroacetate
SMILES C(=O)(C(Cl)(Cl)Cl)OC(=O)C(Cl)(Cl)Cl

Specifications

Color Colorless
Boiling Point 116°C
Quantity 25 g
UN Number 3265
Formula Weight 308.74
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Trichloroacetic Anhydride