accessibility menu, dialog, popup

UserName

CAS RN 4122-04-7

IUPAC Name: 1,3,5-triazin-2-amine
Synonyms: 1,3,5-triazin-2-amine
Molecular Weight (g/mol): 96.09
Molecular Formula: C3H4N4
InChi Key: KCZIUKYAJJEIQG-UHFFFAOYSA-N
SMILES: NC1=NC=NC=N1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results