accessibility menu, dialog, popup

UserName

CAS RN 41105-35-5

Cl

CAS RN 41105-35-5

IUPAC Name: 1-chloro-9,10-bis(2-phenylethynyl)anthracene
Synonyms: 1-chloro-9,10-bis(2-phenylethynyl)anthracene
Molecular Weight (g/mol): 412.92
Molecular Formula: C30H17Cl
InChi Key: IMMCAKJISYGPDQ-UHFFFAOYSA-N
SMILES: ClC1=C2C(=CC=C1)C(C#CC1=CC=CC=C1)=C1C=CC=CC1=C2C#CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results