accessibility menu, dialog, popup

UserName

CAS RN 41078-70-0

IUPAC Name: 3-(2-chloroethyl)-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one
Synonyms: 3-(2-chloroethyl)-2-methylpyrido[1,2-a]pyrimidin-4-one
Molecular Weight (g/mol): 222.67
Molecular Formula: C11H11ClN2O
InChi Key: LFTGLYCNMGGMKL-UHFFFAOYSA-N
SMILES: CC1=C(CCCl)C(=O)N2C=CC=CC2=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results