accessibility menu, dialog, popup

UserName

CH3NBr

CAS RN 41037-28-9

IUPAC Name: 6-bromo-4-methylquinoline
Synonyms: 6-bromo-4-methylquinoline
Molecular Weight (g/mol): 222.09
Molecular Formula: C10H8BrN
InChi Key: XBZQSJLYRMZRSZ-UHFFFAOYSA-N
SMILES: CC1=CC=NC2=CC=C(Br)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results