accessibility menu, dialog, popup

UserName

CAS RN 41031-50-9

IUPAC Name: 2-(adamantan-1-yl)-4-methylphenol
Synonyms: 2-(adamantan-1-yl)-4-methylphenol
Molecular Weight (g/mol): 242.36
Molecular Formula: C17H22O
InChi Key: XHLJIHBDAJFXBE-UHFFFAOYSA-N
SMILES: CC1=CC(=C(O)C=C1)C12CC3CC(CC(C3)C1)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results