accessibility menu, dialog, popup

UserName

HOFFFF

CAS RN 4084-38-2

IUPAC Name: (2,3,5,6-tetrafluorophenyl)methanol
Synonyms: (2,3,5,6-tetrafluorophenyl)methanol
Molecular Weight (g/mol): 180.1
Molecular Formula: C7H4F4O
InChi Key: AGWVQASYTKCTCC-UHFFFAOYSA-N
SMILES: OCC1=C(F)C(F)=CC(F)=C1F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)