accessibility menu, dialog, popup

UserName

ClONH2NOO

CAS RN 40763-96-0

IUPAC Name: 5-chloro-2-nitrobenzamide
Synonyms: 5-chloro-2-nitrobenzamide
Molecular Weight (g/mol): 200.58
Molecular Formula: C7H5ClN2O3
InChi Key: MKHXTOPPKVFSFI-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(C=CC(Cl)=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)