accessibility menu, dialog, popup

UserName

CAS RN 407-38-5

IUPAC Name: 2,2,2-trifluoroethyl 2,2,2-trifluoroacetate
Synonyms: acetic acid,trifluoro, 2,2,2trifluoroethyl ester
Molecular Weight (g/mol): 196.05
Molecular Formula: C4H2F6O2
InChi Key: ZKUJOCJJXCPCFS-UHFFFAOYSA-N
SMILES: FC(F)(F)COC(=O)C(F)(F)F
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)