accessibility menu, dialog, popup

UserName

CH3OOHSOHOOO

CAS RN 4065-45-6

IUPAC Name: 5-benzoyl-4-hydroxy-2-methoxybenzene-1-sulfonic acid
Synonyms: sungard|benzophenone 4|uvinul|sulisobenzone|Uval|uvinul MS 40|spectra-sorb UV 284|sulisobenzona|seesorb 101S|syntase 230
Molecular Weight (g/mol): 308.3
Molecular Formula: C14H12O6S
InChi Key: CXVGEDCSTKKODG-UHFFFAOYSA-N
SMILES: COC1=CC(O)=C(C=C1S(O)(=O)=O)C(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)