CAS RN 40400-13-3
CAS RN 40400-13-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Iodobenzyl Bromide 98.0+%, TCI America™
CAS: 40400-13-3 Molecular Formula: C7H6BrI Molecular Weight (g/mol): 296.933 MDL Number: MFCD00236046 InChI Key: GQFITODJWOIYPF-UHFFFAOYSA-N Synonym: 2-iodobenzyl bromide,1-bromomethyl-2-iodobenzene,o-iodobenzyl bromide,alpha-bromo-2-iodotoluene,benzene, 1-bromomethyl-2-iodo,2-iodobenzylbromide,2-iodo benzylbromide,pubchem14812,acmc-1anor,alpha-bromo-2-iodo toluene PubChem CID: 11426472 IUPAC Name: 1-(bromomethyl)-2-iodobenzene SMILES: C1=CC=C(C(=C1)CBr)I