missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobenzyl Bromide 98.0+%, TCI America™

$44.28 - $127.07

Chemical Identifiers

CAS 40400-13-3
Molecular Formula C7H6BrI
Molecular Weight (g/mol) 296.933
MDL Number MFCD00236046
InChI Key GQFITODJWOIYPF-UHFFFAOYSA-N
Synonym 2-iodobenzyl bromide, 1-bromomethyl-2-iodobenzene, o-iodobenzyl bromide, alpha-bromo-2-iodotoluene, benzene, 1-bromomethyl-2-iodo, 2-iodobenzylbromide, 2-iodo benzylbromide, pubchem14812, acmc-1anor, alpha-bromo-2-iodo toluene
PubChem CID 11426472
IUPAC Name 1-(bromomethyl)-2-iodobenzene
SMILES C1=CC=C(C(=C1)CBr)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I06351G
View Documents
TCI America
I06351G
1 g
Each for $44.28
Only null left
 
I06355G
View Documents
TCI America
I06355G
5 g
Each for $127.07
Only null left
 
Description

Description

Specifications

Chemical Identifiers

40400-13-3
296.933
GQFITODJWOIYPF-UHFFFAOYSA-N
11426472
C1=CC=C(C(=C1)CBr)I
C7H6BrI
MFCD00236046
2-iodobenzyl bromide, 1-bromomethyl-2-iodobenzene, o-iodobenzyl bromide, alpha-bromo-2-iodotoluene, benzene, 1-bromomethyl-2-iodo, 2-iodobenzylbromide, 2-iodo benzylbromide, pubchem14812, acmc-1anor, alpha-bromo-2-iodo toluene
1-(bromomethyl)-2-iodobenzene

Specifications

40400-13-3
125°C
MFCD00236046
3261
GQFITODJWOIYPF-UHFFFAOYSA-N
1-(bromomethyl)-2-iodobenzene
11426472
≥98.0% (GC)
2-Iodobenzyl Bromide
55°C
C7H6BrI
1 g
2-iodobenzyl bromide, 1-bromomethyl-2-iodobenzene, o-iodobenzyl bromide, alpha-bromo-2-iodotoluene, benzene, 1-bromomethyl-2-iodo, 2-iodobenzylbromide, 2-iodo benzylbromide, pubchem14812, acmc-1anor, alpha-bromo-2-iodo toluene
C1=CC=C(C(=C1)CBr)I
296.933
296.93
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'