accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNN

CAS RN 402718-29-0

IUPAC Name: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbonitrile
Synonyms: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbonitrile
Molecular Weight (g/mol): 230.07
Molecular Formula: C12H15BN2O2
InChi Key: BOIKCRIMIQAFQJ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(=CN=C1)C#N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)