accessibility menu, dialog, popup

UserName

CAS RN 39971-65-8

IUPAC Name: zinc(2+) ditrifluoromethanesulfinate
Synonyms: zinc(2+) ditrifluoromethanesulfinate
Molecular Weight (g/mol): 331.51
Molecular Formula: C2F6O4S2Zn
InChi Key: UANOWFITUWBPCF-UHFFFAOYSA-L
SMILES: [Zn++].[O-]S(=O)C(F)(F)F.[O-]S(=O)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results