accessibility menu, dialog, popup

UserName

CAS RN 399-94-0

IUPAC Name: 2-bromo-1,4-difluorobenzene
Synonyms: 2-bromo-1,4-difluorobenzene
Molecular Weight (g/mol): 192.99
Molecular Formula: C6H3BrF2
InChi Key: XCRCSPKQEDMVBO-UHFFFAOYSA-N
SMILES: FC1=CC=C(F)C(Br)=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)