accessibility menu, dialog, popup

UserName

NH2SNClCl

CAS RN 39893-80-6

IUPAC Name: 4-(3,4-dichlorophenyl)-1,3-thiazol-2-amine
Synonyms: 4-(3,4-dichlorophenyl)-1,3-thiazol-2-amine
Molecular Weight (g/mol): 245.12
Molecular Formula: C9H6Cl2N2S
InChi Key: UWAXMUSNRIUHRQ-UHFFFAOYSA-N
SMILES: NC1=NC(=CS1)C1=CC=C(Cl)C(Cl)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results