CAS RN 39890-98-7
CAS RN 39890-98-7
IUPAC Name:
2,6-dichloro-4-(trifluoromethyl)pyridine
Synonyms:
2,6-dichloro-4-(trifluoromethyl)pyridine
Molecular Weight (g/mol):
215.98
Molecular Formula:
C6H2Cl2F3N
InChi Key:
KVNQWVYYVLCZKK-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC(Cl)=NC(Cl)=C1
1
–
1
of
1
results
1
2,6-Dichloro-4-(trifluoromethyl)pyridine 97.0+%, TCI America™
CAS: 39890-98-7 Molecular Formula: C6H2Cl2F3N Molecular Weight (g/mol): 215.98 MDL Number: MFCD00042246 InChI Key: KVNQWVYYVLCZKK-UHFFFAOYSA-N Synonym: 2,6-dichloro-4-trifluoromethyl pyridine,2,6-dichloro-4-trifluoromethyl-pyridine,4-trifluoromethyl-2,6-dichloropyridine,2,6-dichloro-4-trifluoromethylpyridine,acmc-1air6,abbypharma ap-30-1899,pyridine, 2,6-dichloro-4-trifluoromethyl PubChem CID: 520968 IUPAC Name: 2,6-dichloro-4-(trifluoromethyl)pyridine SMILES: FC(F)(F)C1=CC(Cl)=NC(Cl)=C1