CAS RN 39819-65-3
CAS RN 39819-65-3
IUPAC Name:
nickel(2+) dibenzenesulfonate
Synonyms:
nickel(2+) dibenzenesulfonate
Molecular Weight (g/mol):
373.02
Molecular Formula:
C12H10NiO6S2
InChi Key:
IBVWKDVFDAWRFU-UHFFFAOYSA-L
SMILES:
[Ni++].[O-]S(=O)(=O)C1=CC=CC=C1.[O-]S(=O)(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Nickel(II) Benzenesulfonate Hexahydrate 98.0+%, TCI America™
CAS: 39819-65-3 Molecular Formula: C12H10NiO6S2 Molecular Weight (g/mol): 373.019 MDL Number: MFCD00070595 InChI Key: IBVWKDVFDAWRFU-UHFFFAOYSA-L Synonym: Benzenesulfonic Acid Nickel(II) Salt PubChem CID: 14325094 IUPAC Name: benzenesulfonate;nickel(2+) SMILES: C1=CC=C(C=C1)S(=O)(=O)[O-].C1=CC=C(C=C1)S(=O)(=O)[O-].[Ni+2]