accessibility menu, dialog, popup

UserName

CAS RN 39665-12-8

IUPAC Name: (2S)-2,6-diaminohexanoic acid hydrate
Synonyms: lysine monohydrate
Molecular Weight (g/mol): 164.21
Molecular Formula: C6H16N2O3
InChi Key: HZRUTVAFDWTKGD-XZNNNFJINA-N
SMILES: O.NCCCC[C@H](N)C(O)=O
Quantity 
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (5)
  • (2)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (1)
  • (5)
  • (5)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)
  • (2)
  • (5)