accessibility menu, dialog, popup

UserName

H3COClO

CAS RN 39649-71-3

IUPAC Name: 4-(hexyloxy)benzoyl chloride
Synonyms: 4-(hexyloxy)benzoyl chloride
Molecular Weight (g/mol): 240.73
Molecular Formula: C13H17ClO2
InChi Key: DQQOONVCLQZWOY-UHFFFAOYSA-N
SMILES: CCCCCCOC1=CC=C(C=C1)C(Cl)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)