accessibility menu, dialog, popup

UserName

CAS RN 39637-74-6

IUPAC Name: (1S,4R)-4,7,7-trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carbonyl chloride
Synonyms: (1S,4R)-4,7,7-trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carbonyl chloride
Molecular Weight (g/mol): 216.66
Molecular Formula: C10H13ClO3
InChi Key: PAXWODJTHKJQDZ-VHSXEESVSA-N
SMILES: CC1(C)[C@@]2(C)CC[C@@]1(OC2=O)C(Cl)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)