accessibility menu, dialog, popup

UserName

H3COOCH3NCS

CAS RN 39574-16-8

IUPAC Name: ethyl 2-isothiocyanatopropanoate
Synonyms: ethyl 2-isothiocyanatopropanoate
Molecular Weight (g/mol): 159.2
Molecular Formula: C6H9NO2S
InChi Key: ALJGYASQFZQQJX-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C)N=C=S
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)