accessibility menu, dialog, popup

UserName

CH3OOOHOH3C

CAS RN 3943-74-6

IUPAC Name: methyl 4-hydroxy-3-methoxybenzoate
Synonyms: vanillate methyl vanillate
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: BVWTXUYLKBHMOX-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(O)C(OC)=C1
Boiling Point 
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)